VWR®, Stem Thermometer, Calibrated, Electronic, Stainless Steel, 120 mm Probe
Lieferant: VWR Collection
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
620-2200EA 68.8 EUR InStock
620-2200
VWR®, Stem Thermometer, Calibrated, Electronic, Stainless Steel, 120 mm Probe
Thermometer Digitalthermometer
Digital stem thermometer suitable for field studies, wet lab applications, wash-down areas, food testing and tanks.
  • Will read temperatures in liquids, semi-solids and gases
  • Stainless steel probe easily reaches into cuvettes, test tubes, flasks, beakers and more
  • Includes accredited calibration documentation that meets the requirements of ISO 17025
  • Individually serialised
Zertifizierungen: Tested and calibrated in exclusive triple-accredited/registered ISO/IEC 17025:2017, A2LA accredited laboratory (Cert #2448,01), against equipment whose calibration is traceable to NIST.
Lieferumfang: Includes multi-language instructions (English, French, German, Italian, Portuguese and Spanish).
Achtung! Bitte beachten: These electronic stem thermometers are not suitable for solids such as frozen items.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge