VWR® AM 100, Raumklima-Monitor
Lieferant: VWR Collection
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
620-2624EA 201 EUR InStock
620-2624
VWR® AM 100, Raumklima-Monitor
Überwachungssysteme CO2- und Temperatur-Überwachungssysteme
Gleichzeitige Messung und Anzeige von Lufttemperatur, Luftfeuchtigkeit und CO₂-Konzentration.
  • Three different LEDs (green, yellow, red) indicate the CO₂ value in the air as good, medium or bad
  • Acoustic alarm when CO₂ limit is exceeded
  • Helps to improve room climate and prevent mould formation
  • Large LCD with backlight for easy reading in the dark
  • MUTE button for alarm on/off
  • For bench top or wall mounting
This smart, compact, easy to use device was designed to help people taking care of Indoor Air Quality. The AM 100 can be widely used in office buildings, schools, exhibition halls, shopping malls, meeting rooms, fitness centres, restaurants and other public places where personal comfort and a healthy environment is important.
Lieferumfang: Wird mit AC/DC-Adapter und Werkskalibrierzertifikat geliefert.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge