VWR® Traceable® Digital Heat Thermometer
Lieferant: VWR Collection
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
620-2809EA 51.4 EUR InStock
620-2809
VWR® Traceable® Digital Heat Thermometer
Thermometer Digitalthermometer
Simultaneously displays temperature, timer, and current alarms.
  • Tri-display temperature, alarm, timer
  • Programmable high/low alarm
  • ºC/ºF exchange
  • 12/24 hour countdown timer
  • Timer setting memory
  • Timer elapse time display
  • Stainless steel probe
  • Magnetic attachment
  • Low battery indicator
Perfect for use in ovens and incubators.
Note: New product design for 620-1912.
Zertifizierungen: Individually calibrated ISO 17025 certified. Individually certified—traceable to NIST.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge