Plastic refrigerator/freezer thermometer
Lieferant: TFA DOSTMANN
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
144.006
620-8204EA 4.1 EUR InStock
620-8204
Plastic refrigerator/freezer thermometer
Thermometer Analoge Thermometer
This useful plastic thermometer shows you at a glance whether vegetables, cheese, meat, etc. are properly cooled.
  • You can easily hang in your fridge or freezer with the handy mounting part
  • Temperature is measured with proven bimetallic technology
  • With freezing-zones indication for easy reading
The product is entirely manufactured in Germany.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge