62108.03
62108.03EA
134
EUR
InStock
62108.03
Decacarbonyldirhenium ≥96%
Decacarbonyldirhenium
Purity: 96%
CAS No.: 14285-68-8
Molecular Formula: C10O10Re2
Formula Weight: 652.52
Physical Form: Powder
Density: 2.87
Appearance: White
Melting Point: 170° dec.
UN No.: 3466
MDL No.: MFCD00011198