Eyepiece cameras, Dino-Eye
Lieferant: Dino-Lite / IDCP B.V.
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
AM4023 AM4023CT AM4023U AM7025X
630-1472EA 0 EUR InStock
630-1472 630-1473 630-1474 630-2922
Eyepiece cameras, Dino-Eye
Kameras
With these cameras users can turn their conventional microscopes into digital microscopes.The ocular (eyepiece) of the non-digital microscope is simply replaced by an Dino-Eye eyepiece and connected to a computer via USB.
  • Different models for different eyepiece sizes
  • High speed USB 2.0 connection
  • Software included for image or video processing, incl. calibrated measurement features
  • MicroTouch feature (model AM4023CT only): Touch sensitive area that users can use to capture images instead of using a keyboard and mouse
  • Model AM7025X: Edge series with improved colour accuracy and larger field of view
  • Models AM4023CT and AM7025X also connect to devices with C-mount
Edge model is suitable for demanding applications where true colour reproduction is of the utmost importance, like with pathology samples or quality control.
Lieferumfang: Supplied with carrying pouch, software CD (DinoCapture (Windows) and DinoXcope (MAC)), calibration target and user manual.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge