Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Handmikroskope, Dino-Lite® EarScope

Lieferant: Dino-Lite / IDCP B.V.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

MEDL3E MEDL4E MEDL4EP
630-2607EA 249 EUR InStock
630-2607 630-2609 630-2610
Handmikroskope, Dino-Lite® EarScope
Mikroskope
With this handheld microscope users can easily inspect and photograph the ear canal, the ear drum and the outer ear. Ideal for use in general practice, in Audiology and for ear, nose and throat specialists.

  • Help to identify ear problems quickly
  • Bright and clear image thanks to built-in LED lights
  • High speed USB 2.0 connection
  • Speculum is removable and can be cleaned or replaced

The image can be viewed on a computer screen to simplify diagnosis simplifies and also allows the patient to watch the images as well. The images, either still photo or video, can be stored digitally and easily added to a patient file or forwarded to a colleague.

The EarScope Basic MEDL3E has a VGA resolution and is particularly suitable for home use. The speculum is removable and can be cleaned.

The EarScope Pro MEDL4E is intended for professional use, has a 1,3 MP resolution and comes with replaceable specula of 3, 4 and 5 mm. Ten pieces of each size are included and re-ordering of the specula is possible.

The EarScope Pneumatic MEDL4EP is the top model with a 1,3 MP resolution. With this model, the physician has the additional function of blowing a puff of air into the ear canal which allows him to assess the mobility of the eardrum that can point to a glue ear. Just like the EarScope MEDL4E this model has replaceable specula, in this case 2,5 and 4 mm.

Zertifizierungen: Entspricht der Medizinprodukterichtlinie 93/42/EWG mit Ergänzung 2007/47/EG.

Lieferumfang: Wird mit eine Tasche, einer Software-CD und einem Kalibrierblatt geliefert.

Order Now
 

PRODUKT

TECHNISCHE DATEN

ZUBEHÖR

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge