Handheld digital microscope, Dino-Lite Medical, Podoscope
Lieferant: Dino-Lite / IDCP B.V.
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
MEDL4FW
630-2611EA 559 EUR InStock
630-2611
Handheld digital microscope, Dino-Lite Medical, Podoscope
Mikroskope
With this handheld microscope changes in the skin of the foot and the foot itself can be documented by taking photographs and making movies as well. The podiatrist and his patient can view the images in real time on a laptop, Windows tablet or computer.
  • Versatility due to exchangeable caps
  • Built-in lighting (eight bright white LEDs)
  • High speed USB 2.0 connection
  • Powered via USB port: No batteries needed
  • Built-in anti-reflection polariser will help to reduce any bothersome shine or glow of the skin
Designed for use by podiatrists.
Zertifizierungen: Certified according to the Medical Device Directive 93/42/EEC, modified 2007/47/EC.
Lieferumfang: Supplied with carry pouch, software CD (DinoCapture 2.0 (Windows), DinoXcope (Mac OS)), calibration target and medical user manual.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge