Handmikroskope, DermaScope®
Lieferant: Dino-Lite / IDCP B.V.
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
MEDL7DM MEDL4DW MEDL7DW
630-2617EA 859 EUR InStock
630-2617 630-2511 IDCPMEDL7DW
Handmikroskope, DermaScope®
Mikroskope
These handy, easy-to-use microscopes can be quickly deployed and used to create sharp and clear photos and videos of a wide variety of skin problems.
  • Without pain or long waiting times for the patient
  • Built-in adjustable polariser reduces the gloss effects of the skin
  • Support the use of water or oil (not always necessary)
  • Illumination: 8 white LEDs (switchable)
  • High speed USB 2.0 output
  • HR models come with a metal housing
  • Wireless streaming to a tablet, smartphone, or computer is possible in combination with WF-10 unit (to be ordered separately)
Ideal for physicians, dermatologists or skin specialists.
Zertifizierungen: Entspricht der Medizinprodukterichtlinie 93/42/EWG mit Ergänzung 2007/47/EG.
Lieferumfang: Wird mit einer Tasche, einer Software-CD und einem Kalibrierblatt geliefert.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge