VWR® VisiCam® TC 20 Plus, Tablet-Kamera, drahtlos
Lieferant: VWR Collection
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
630-2661EA 1020 EUR InStock
630-2661
VWR® VisiCam® TC 20 Plus, Tablet-Kamera, drahtlos
Kameras
This system is composed of a tablet and a wireless 5 MP camera. It can be attached to all microscopes with a C-mount.
  • No need for a computer
  • Wireless connection between tablet and camera: Tablet can be used in any position
  • Images can be shared immediately through wireless network
  • Tablet with user friendly 10,1" touch screen display
  • Quick and easy creation of data reports
Image analysis software pre-installed on the tablet has been developed so that user experience is easy and intuitive. The software allows preview, image capture, video recording, measuring, annotations, image parameters adjustment, data export to PDF or Excel, counting, multi-user password protected, quick sharing of images and reports.
Tablet specifications:
Operating system: Android OS
CPU: Processor Intel® Atom™ Z3745, 1,33 GHz, Quad Core
RAM: 1024 MB
Display: LCD high definition 10,1", multi-touch
Memory: HD SSD 16 GB
Network: Wireless, Bluetooth
Bestellinformation: Included in the package: 10" tablet supplied with multilingual software for image capture and analysis (EN, FR, DE, PT, ES, IT), Lithium polymer (LiPo) eight hour lifetime battery, wireless CMOS camera, power supply, slide with calibration circles, and table support.
Lieferumfang: Lieferung mit mehrsprachiger Software für Bilderfassung und -analyse, Netzteil (mit EU-, UK- und US-Stecker), Kalibrierungsobjektträger und Tischhalterung für das Tablet.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge