Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Routine stereo microscope, M60

Lieferant: LEICA MICROSYSTEMS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

12730528 10450294_P
630-3330EA 472 EUR InStock
630-3330 630-3159
Routine stereo microscope, M60
Mikroskope
Easily inspect, screen, observe or document.

  • Customise this microscope to suit your needs with a large range of ergonomic accessories and illumination adapters
  • Increase your productivity with an entirely ergonomic microscope workplace to work comfortably
  • Create traceable measurement procedures for repetitive jobs using the M60's click stops

Handle your daily inspection or documentation, work confidently, and stay flexible in times of change. The modular stereo microscopes of the M series from Leica are the right choice for an abundant variety of tasks in life sciences and industrial applications.

The M60 allows operators to see a large sample overview, work comfortably under the microscope, and capture images of important details easily.

M series stereo microscopes have been designed to provide tailored solutions for operators working in laboratories or in manufacturing. Enjoy working with Leica optics that deliver outstanding resolution paired with great depth of field, so that you see all relevant structures at a glance.

You can easily change from sample overview to fine detail imaging. No refocusing is needed when changing magnification - parfocal optics allow you to keep your sample in focus at all zoom levels.

M60 with 6:1-zoom range has a continuous zoom from 6,3× to 40× including engageable click stops - making repetitive inspection and measurement at a specific zoom position reliable. This system is optimised for a larger field of view to see more at a glance.

Zubehörinformationen: Leica IC90 E camera (sold separately) includes a USB cable, type A-B, and CD with Leica Application Suite (LAS) software.

The standalone kit (sold separately) enables you to connect your Leica IC90 E to a HDMI monitor and store images on a SD card. Recommended if you use the camera without a PC. Contents: HDMI cable, type A-A, Remote control RC3 (model 10 450 805), SD card, Power supply with international connectors for US, EU, UK, and AU, and Ethernet cable.

AirTeach software (sold separately) is compatible with the following cameras and microscopes: ICC50W&E, EZ4W&E, S9i, and IC90E.

Bestellinformation: 2 year warranty.

Lieferumfang: Package includes: Leica M60 optics carrier, two eyepieces (10×/23B, adjustable), ErgoTube 45°, objective (achromat, 0.8×, WD = 114 mm), Leica LED2000, power cable (2 m, EU plug), dust cover (antistatic, small).

Order Now
 

PRODUKT

TECHNISCHE DATEN

ZUBEHÖR

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge