Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Imager Tissue Package, THUNDER

Lieferant: LEICA MICROSYSTEMS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

11510061_P1
630-3395EA 0 EUR InStock
630-3395
Imager Tissue Package, THUNDER
Mikroskope
The THUNDER imager tissue allows real time fluorescence imaging of 3-D tissue sections typically used in neuroscience and histology research. Acquire rich, detailed images of thick tissues free of haze from ‘out of focus’ blur.

Zertifizierungen: CE and CAS.

Lieferumfang: THUNDER Imager Tissue Imager Package includes: DM4 B semi-motorised upright microscope stand, manual camera port, motorised 5-position fluorescence filter turret, coded 7-position objective nosepiece, objectives (Plan FL 1,25×/0,04 N.A., Plan FL 10×/0,32 N.A. and Plan Apo 40×/0,95 N.A. with correction collar), 10×/25 mm FOV eyepieces, LED transmitted light, brightfield condenser (0,90 N.A.), manual stage (110° rotation), shutter controlled LED3 fluorescence light source, fluorescence filters (LED405, GFP and RHOD), K5 sCMOS camera; dust cover, THUNDER Workstation 2D, US keyboard, LAS X Core software, LAS X Multi-Channel Acquisition software module, 32" monitor and 1 TB SSD RAID addition.
Purchase of THUNDER Imager Tissue Package includes service installation and 1 day of applications training by an LMS Advanced Workflow Specialist.

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge