VWR® Polysine®, Adhäsionsobjektträger,
Lieferant: VWR Collection
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
631-0107EA 34.1 EUR InStock
631-0107
VWR® Polysine®, Adhäsionsobjektträger,
Objektträger Adhäsions-Objektträger
These slides are coated with a permanent adhesive that provides excellent cell and tissue adhesion.
  • Tissue section thickness of 2 to 5 µm
  • 90° ground edges and colour-frosted end for labelling
  • Adhesion superior to glue-, protein- or silane-treated slides
Recommended for molecular biology techniques and cytocentrifuged preparations to reduce tissue loss during prolonged DNA hybridisation and immunocytochemistry.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge