Deckgläser, rund
Lieferant: MARIENFELD
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
0111540 0111530 0111700 0111580 0111560 0111600 0111620 0111640 0111500 0111520 0111550 0111650
631-0692EA 66.9 EUR InStock
631-0692 630-1597 631-0694 631-0669 631-0668 631-0670 631-0671 631-0672 631-0665 631-0666 631-0667 MARI0111650
Deckgläser, rund
Deckgläser
Round cover glasses made of pure white borosilicate D 263 ™ M glass, Hydrolytic Class I.
  • No bubbles or flaws
Suitable for in vitro diagnostic (IVD) applications.
Verpackung: 100 cover glasses per box with separate lid.
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge