Tissue embedding sponge, biopsy test
Lieferant: DELTALAB
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
192922.04
631-9681EA 73.8 EUR InStock
631-9681
Tissue embedding sponge, biopsy test
Biopsieschwämmchen
Foam sponge, very useful for small specimens in biopsy tests.
  • Avoids sample loss though drain holes when sample is processed
  • Does not affect parafin drainage
Fits standard cassettes.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge