Stak-a-Tray system, Bel-Art ProCulture, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
H186100470 H186101440
631-9775EA 253 EUR InStock
631-9775 BELAH186101440
Stak-a-Tray system, Bel-Art ProCulture, SP Bel-Art
Gestelle Inkubatorgestelle
These are stainless steel modular rack and tray system that maximizes storage for petri dishes and cell culture flasks in incubators, cold rooms or on the bench.
  • Perforated trays allow air flow and slide easily in the rack
  • Stack up to five racks for 20 levels of storage
  • Each tier accommodates one large or two small trays
All parts are stainless steel and autoclavable at 121 ºC (250 ºF).
Bestellinformation: Racks and trays are sold separately.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge