Economical Storage Box
Lieferant: DELTALAB
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
DS42
631-9790EA 457 EUR InStock
631-9790
Economical Storage Box
Boxen Lagerboxen
Made of heavy-duty cardboard for long term storage.
This box, complete with a lid, is specially designed for the storage of histology cassettes.
Each box stores up to 250 cassettes or 160 embedding rings.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge