Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

D(+)-Galactose ≥99% (durch HPLC), Multikompendial endotoxinarm, J.T.Baker®

Lieferant: AVANTOR PERFORMANCE MATERIALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


6322-05EA 1112.2 EUR InStock
6322-05 6322-06 6322-07
D(+)-Galactose ≥99% (durch HPLC), Multikompendial endotoxinarm, J.T.Baker®
D(+)-Galactose


Formel: C₆H₁₂O₆
MW: 180.16 g/mol
Siedepunkt: 527
Dichte: 1.72 g/cm³
MDL Number: MFCD00151230
CAS-Nummer: 59-23-4
Merck Index: 13,04356
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=508b1d21a6801ddfb1a5f13a64cb06e6f95e8922&vid=508b1d21a6801ddfb1a5f13a64cb06e6f95e8922&prid=web&clid=SAPDAM
Order Now
Spezifikation Testergebnisse
Meets B.P. Chemical Specifications
GMP Manufactured Product
Meets E.P. Chemical Specifications
Preserve in well-closed containers.
CAUTION: For Manufacturing, processing or repackaging Passes Test
Bulk Pharmaceutical Chemical Passes Test
NF - Assay (C6H12O6 ) (anhydrous basis) 98.0 - 102.0 %
NF - Appearance of Solution Passes Test
NF - Acidity Passes Test
NF - Identification A (IR) Passes Test
NF - Identification B (HPLC) Passes Test
NF - Identification C Passes Test
NF - Optical Rotation [Å]²ºD (+) 78.0 - 81.5 Degree
NF - Related Substances: Lactose and 1,6-galactosylgalactose ≤ 0.6 %
NF - Related Substances: Galacturonic acid ≤ 0.6 %
NF - Related Substances: Dextrose ≤ 0.6 %
NF - Related Substances: Tagatose ≤ 0.6 %
NF - Related Substances: Dulcitol ≤ 0.6 %
NF - Related Substances: Arabinose ≤ 0.6 %
NF - Related Substances: Any unspecified impurity, each ≤ 0.2 %
NF - Related Substances: Total impurities ≤ 1.0 %
NF - Barium (Ba) Passes Test
NF - Lead (Pb) ≤ 0.5 ppm
NF - Water (H₂O) ≤ 1.0 %
NF - Residue on Ignition ≤ 0.1 %
EP/BP - Assay (C₆H₁₂O₆)(anhydrous basis) 97.0 - 102.0 %
EP/BP - Appearance of Solution Passes Test
EP/BP - Acidity or Alkalinity Passes Test
EP/BP - Identification A (IR) Passes Test
EP/BP - Proteins ≤ 0.1 %
EP/BP - Related Substances: Sum of Impurities A and B ≤ 1.0 %
EP/BP - Related Substances:Unspecified impurities, each ≤ 0.3 %
EP/BP - Related Substances:Total impurities ≤ 2.0 %
EP/BP - Specific Optical Rotation [Å]²ºD (+) 78.0 - 81.5 Degree
EP/BP - Water (H₂O) ≤ 1.0 %
EP/BP - Sulfated Ash ≤ 0.1 %
EP/BP - TAMC (cfu/g) ≤ 100 cfu/g
Appearance Passes Test
Endotoxin Concentration (EU/g) ≤ 2.5
Glucose ≤ 0.1 %
Purity (HPLC) ≥ 99 %
Residue on Ignition ≤ 0.05 %
Microbial Testing - Salmonella (USP) None Detected
Microbial Testing - E. coli (USP) None Detected
Microbial Testing - Staphylococcus aureus (USP) None Detected
Microbial Testing - Pseudomonas aeruginosa (USP) None Detected
Microbial Testing - Total Yeast and Mold (USP) ≤ 100 cfu/g
Microbial Testing - Total Aerobic Microbial Count (USP) ≤ 100 cfu/g
Solution Clarity (300 mg/mL) Passes Test
Solution Color, APHA - For Information Only For Information Only
Trace Impurities - Aluminum (Al), For Information Only For Information Only
Trace Impurities - Arsenic (As), For Information Only For Information Only
Trace Impurities - Cadmium (Cd), For Information Only For Information Only
Trace Impurities - Chromium (Cr), For Information Only For Information Only
Trace Impurities - Copper (Cu), For Information Only For Information Only
Trace Impurities - Iridium (Ir), For Information Only For Information Only
Trace Impurities - Iron (Fe), For Information Only For Information Only
Trace Impurities - Lead (Pb), For Information Only For Information Only
Trace Impurities - Manganese (Mn), For Information Only For Information Only
Trace Impurities - Mercury (Hg), For Information Only = 3000 ppm
Trace Impurities - Molybdenum (Mo), For Information Only = 5000 ppm
Trace Impurities - Nickel (Ni), For Information Only = 5000 ppm
Trace Impurities - Osmium (Os), For Information Only
Trace Impurities - Palladium (Pd), For Information Only
Trace Impurities - Platinum (Pt), For Information Only
Trace Impurities - Rhodium (Rh), For Information Only
Trace Impurities - Ruthenium (Ru), For Information Only
Trace Impurities - Vanadium (V), For Information Only
Trace Impurities - Zinc (Zn), For Information Only
Residual Methanol ≤ 3000 ppm
Residual Ethanol ≤ 5000 ppm
Residual Isopropanol ≤ 5000 ppm
Residual Solvents: Only the Class 2 solvent Methanol and the Class 3
Solvent Ethanol are likely to be present. Each is tested and the
concentration reported for each batch.
Metallic Residues: No metal catalysts or metal reagents, as defined by
EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of
this material.
Plant Derived

 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge

Diese Webseite verwendet Cookies, eigene oder von Drittanbieter, zur Sammlung statistischer Informationen, um Inhalte und Anzeigen für Sie zu personalisieren. Wenn Sie diese Seite weiterhin verwenden, akzeptieren Sie die Cookie-Nutzung.

Weitere Informationen finden Sie in unseren Datenschutzbestimmungen..