Reflectance colorimeters, RT 300, RT 400, RT 500, Lovibond®
Lieferant: Lovibond Tintometer
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
400300 400400 400500 400310
633-0247EA 10050 EUR InStock
633-0247 633-0249 633-0250 633-0248
Reflectance colorimeters, RT 300, RT 400, RT 500, Lovibond®
Kolorimeter Reflexions-Kolorimeter
The RT300, RT400 and RT500 are versatile, portable spectrophotometers in a choice of diffuse/8° sphere or 0/45° optical geometry that are designed to give fast, precise and accurate colour measurement information on a range of products. They provide absolute and difference measurements in various colorimetric systems.
The RT300 and RT500 can be combined with the OnColor™ software package allowing measurement data to be downloaded, analysed in detail and stored indefinitely on a PC.
  • Lightweight, compact portable instruments
  • Opacity and colour strength measurement
  • Data storage for 1024 standards with tolerances and 2000 samples
  • Flip-back target shoe for flexible use
  • Rechargeable battery for remote use
Lieferumfang: Supplied with calibration standards, AC adapter, carrying case and operation manual. Other models are available, please contact your local Avantor Sales Office for further details.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge