Handheld refractometer, Acidity meter for vinegar, PAL- Easy ACID 181 master kit
Lieferant: ATAGO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
7781
635-0833EA 390 EUR InStock
635-0833
Handheld refractometer, Acidity meter for vinegar, PAL- Easy ACID 181 master kit
Refraktometer Handrefraktometer
This Master Kit is specifically for measuring vinegar and includes everything you need for easy, precise and accurate measurements.
  • ATAGO's durable titan electrode will show the result that correlated with that of titaration
To measure acidity, sample needs to be dilulted.
Bestellinformation: Requires 2 x AAA alkaline batteries.
Lieferumfang: The master kit comes complete with a scale, 1 beaker, and 1 measuring spoon.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge