Mouse IgG2b Isotype Control [clone: MPC-11] (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®)
Lieferant: Tonbo Biosciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
65-4732-U100
65-4732-U100EA 143 EUR InStock
65-4732-U100
Mouse IgG2b Isotype Control [clone: MPC-11] (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®)
Antikörper
The MPC-11 immunoglobulin is useful as an isotype-matched control. The MPC-11 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG2b antibodies.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge