Armenian Hamster IgG Isotype Control [clone: PIP] (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®)
Lieferant: Tonbo Biosciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
65-4888-U100
65-4888-U100EA 222 EUR InStock
65-4888-U100
Armenian Hamster IgG Isotype Control [clone: PIP] (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®)
Antikörper
The PIP immunoglobulin is useful as an isotype-matched control. The PIP immunolglobulin is specific for Glutathione S-transferase and can be used as an isotype control for Armenian Hamster IgG antibodies.
Type: Control
Antigen: Isotype Control
Clonality: Monoclonal
Clone: PIP
Conjugation: PerCP-Cy5.5
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: N/A
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge