Toluol 99,9, CHROMASOLV™ Plus HPLC, Riedel-de Haën™
Lieferant: HONEYWELL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
Gefahr
650579-1L
650579-1LEA 95.37 EUR InStock
650579-1L
Toluol 99,9, CHROMASOLV™ Plus HPLC, Riedel-de Haën™
Toluol
  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities
Formel: C₆H₅CH₃
MW: 92.14 g/mol
Siedepunkt: 110.6 °C
Melting Pt: –95 °C
Dichte: 0.867 g/cm³ (20 °C)
Flash-Pt: 7.1 %(V)
Storage Temperature: Raumtemperatur
MDL Number: MFCD00008512
CAS-Nummer: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3 II
REACH: 05-2114615130-69
Merck Index: 13,09607
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=7b8a82c36ede4fc1e8822a573f5ce08743569a24&prid=web&clid=SAPDAM
Order Now
Spezifikation Testergebnisse
IR spectrum complying
Assay (GC) Min. 99.90 %
GC (as heptachlor epoxide) Max. 10 ng/l
Non-volatile matter Max. 0.0001 %
Water (Karl Fischer) Max. 0.02 %
Absorbance at 284 nm Max. 1.0
Absorbance at 300 nm Max. 0.10
Absorbance at 325 nm Max. 0.02
Absorbance at 350 nm Max. 0.005
Absorbance at 400 nm Max. 0.005
Autoignition Temperature 997 °F
Vapor Pressure 22 mmHg ( 20 °C)
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge