Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Magnesiumsulfat Heptahydrat USP, Ph. Eur., BP

Lieferant: VWR Chemicals

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


6527.1000EA 58.8 EUR InStock
6527.1000 6527.5000 6527.9025
Magnesiumsulfat Heptahydrat USP, Ph. Eur., BP
Magnesiumsulfat Heptahydrat
Appearance: White or almost white, crystalline powder or brilliant, colourless crystals.

Achtung! Bitte beachten: Not for use as Active Pharmaceutical Ingredient (API)


Formel: MgSO₄·7H₂O
MW: 246.48 g/mol
Melting Pt: 1124 °C
Dichte: 2.66 g/cm³ (20 °C)
Storage Temperature: Raumtemperatur
MDL Number: MFCD00149785
CAS-Nummer: 10034-99-8
EINECS: 231-298-2
Merck Index: 13,05715
Order Now
Spezifikation Testergebnisse
Appearance Conforms (see CoA/CoS)
Heavy metals (as Pb) ≤ 10 ppm
Residual solvents Passes test
Residual metal catalysts (skip lot): .
As (Arsenic) ≤ 2 ppm
Cd (Cadmium) ≤ 0.1 ppm
Co (Cobalt) ≤ 1 ppm
Hg (Mercury) ≤ 2 ppm
Ni (Nickel) ≤ 6 ppm
Pb (Lead) ≤ 2 ppm
V (Vanadium) ≤ 10 ppm
Conforms to Ph.Eur./USP/BP Passes test
Ph.Eur./BP specifications: .
Assay (dried substance) [Ph.Eur./BP] 99.0 - 100.5 %
Identification A Passes test USP
Identification B Passes test USP
Appearance of solution Passes test Ph.Eur./BP
Acidity or alkalinity Passes test Ph.Eur./BP
Cl (Chloride) [Ph.Eur./BP] ≤ 300 ppm
Fe (Iron) [Ph.Eur./BP] ≤ 20 ppm
Loss on drying (400°C) [Ph.Eur./BP] 48.0 - 52.0 %
USP specifications: .
Assay (by ignition) [USP] 98.0 - 102.0 %
Cl (Chloride) [USP] ≤ 0.014 %
Fe (Iron) [USP] ≤ 20 μg/g
Se (Selenium) [USP] ≤ 30 μg/g
pH (25°C; 50 mg/ml) [USP] 5.0 - 9.2
Loss on ignition (450 ± 25°C) [USP] 40.0 - 52.0 %

 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge