Nitrification inhibitor for BOD
Lieferant: HACH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
253335 45901 253334 257924
662-0006EA 54.4 EUR InStock
662-0006 HACH45901 HACH253334 HACH257924
Nitrification inhibitor for BOD
Standards Umweltanalyse-Standards
TCMP - 2-chloro-6 (trichloromethyl) pyridine (N-Serve), coated on an inert substrate.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge