pH electrode calibration and maintenance solutions kit
Lieferant: WTW
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
109415
662-0322EA 116 EUR InStock
662-0322
pH electrode calibration and maintenance solutions kit
Puffer pH-Puffer
Adapted to different electrode types for calibration and maintenance.
The pepsin cleaning solution is used to remove protein containing precipitation from diaphragms, the 3 molar KCl as a storage or refill solution.
Lieferumfang: Kit contains set of 250 ml each of pepsin cleaning solution; 3 M KCl solution and standard buffer solutions of pH 4,006, 6,865, 9,180.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge