ORP-Elektrode, SenTix® ORP-T 900, IDS
Lieferant: WTW
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
103791
662-2020EA 296 EUR InStock
662-2020
ORP-Elektrode, SenTix® ORP-T 900, IDS
Elektroden Redox-Elektroden
IDS ORP electrodes are used to measure oxidation or reduction potentials in aqueous solutions - with integrated temperature sensor.
  • Safe measured values due to signal processing in the sensor
  • Integrated temperature sensor NTC 30 kOhm for precise value documentation
  • Best possible GLP support by documenting the sensor data
Platinum IDS ORP electrode with liquid electrolyte and glass shaft, temperature sensor, ceramic junction.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge