pH/mV/°C/ISE meter, handheld, Eutech pH 450
Lieferant: EUTECH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
ECPHWP45002K ECPHWP45003K ECPHWP45000 ECPHWP45002
662-2275EA 669 EUR InStock
662-2275 662-2312 662-2301 662-2274
pH/mV/°C/ISE meter, handheld, Eutech pH 450
Elektrochemische Messgeräte pH- und Multi-Parameter-Messgeräte
This portable meter is ideal for use in any environment. It features a durable waterproof and dustproof design with IP67 rating.
  • Large, backlit display with intuitive user-friendly icons
  • Grip-Clip™: Quickly and easily secures the pH electrode to a beaker or container
  • Automatic buffer recognition; up to 5- or 6-point calibration; calibration due alarm
  • Stores up to 500 data sets; easy USB or RS232 output with data cables (cables not always included)
  • Meets GLP standards: Built-in real-time clock stamps stored data and calibration data with time and date
  • Can be used with two AA batteries or with optional universal power adapter (not included)
  • Built-in stand with wall-mount option allows for easy bench top measurement or storage
The meter can be used with pH, ORP or ISE electrodes with BNC connector.
Lieferumfang: Meter only (without electrodes) is supplied with Grip-Clip™ and two AA batteries.
Set 1 additionally includes a double junction pH electrode and ATC probe.
Set 2 additionally includes a double-junction pH electrode and ATC probe, hard carry case, USB cable, pH buffers, storage solution and rinse solution.
Set 3 additionally includes a double-junction pH/ATC electrode, hard carry case, USB cable, pH buffers, storage solution and rinse solution.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge