Drucken
pH/mV/°C meters, handheld, Eutech pH 5+ / pH 6+
Lieferant: EUTECH
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
ECPH501PLUS ECPH502PLUSK ECPH503PLUSK ECPH601PLUS ECPH602PLUSK ECPH603PLUSK ECPH601PLUSK
EUTEECPH501PLUSEA 334 EUR
EUTEECPH501PLUS EUTEECPH502PLUSK EUTEECPH503PLUSK EUTEECPH601PLUS 662-2286 662-2287 EUTEECPH601PLUSK
pH/mV/°C meters, handheld, Eutech pH 5+ / pH 6+
Elektrochemische Messgeräte pH- und Multi-Parameter-Messgeräte
These portable meters either measure pH and temperature (pH 5+) or pH, temperature and ORP values (pH 6+). With their compact and sturdy design, the lightweight meters are the ideal choice for field and lab work.
  • Multi-point calibration (up to 5 points) with auto-buffer recognition - choice of USA, NIST or pure water buffer standards with ±0,01 pH accuracy
  • Automatic Temperature Compensation (ATC) for the highest accuracy under fluctuating temperatures
  • Self-diagnostic messages for easy trouble-shooting
  • Shock-resistant and splash resistant ABS plastic casing, rubber boot protects against knocks and hits
  • HOLD function, automatic power-off after 20 minutes of inactivity
  • Convenient benchtop stand, tactile keypad and customised LCD screen
Typical applications include: Pools and spas, ponds and aquariums, agriculture and hydroponics, water and wastewater treatment, cooling towers, food processing, photo-development and printing, ecological studies, field, laboratory and education institutions.