Ion selective electrode, High Performance, ammonia, Orion™
Lieferant: Thermo Orion
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
9512HPBNWP
662-2305EA 773 EUR InStock
662-2305
Ion selective electrode, High Performance, ammonia, Orion™
Elektroden ISE-Elektroden
The high performance ammonia ion selective electrode (ISE) allows fast, simple, economical, and accurate measurements of dissolved ammonia in aqueous solutions.
  • Response time of 1 minute in sample concentrations of 1 ppm and higher; fast results make it easy to combine functionality with autosamplers
  • Keying feature on the electrode body prevents membrane from twisting or breaking during assembly
  • Choice of preassembled outer body with membrane cap or loose membranes
  • New wastewater applications developed using known addition and direct measurement techniques
  • EPA-compliant method
  • Waterproof BNC connector
This gas sensing electrode can also be used to measure the ammonium ion after conversion to ammonia or to measure organic nitrogen after Kjeldahl digestion of the sample. Sample colour and turbidity do not affect the measurement and samples do not need to be distilled. Almost all anions, cations and dissolved species, other than volatile amines, do not interfere.
Lieferumfang: Supplied with 1 m cable, 20 loose membranes, one preassembled outer body and membrane cap, two 60 ml bottles of electrode filling solution, spout cap and tweezers.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge