Multiparameter photometer, Water-ID® The PrimeLab 2.0
Lieferant: WATER ID PRIMELAB
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
PL02BALL PL02BALL-WO90 PL02BALL-COD ELECHRGLATCKIT ELECHRPCATCKIT ELECLRGL-KIT ELECLRPCATCKIT ELORPPCATC-KIT ELPHGLATC-KIT ELPHGLREFATCKT ELPHPCATC-KIT ELECHRGLATC ELECHRPCATC PL2SP-ELECLRGL ELECLRPCATC ELORPPCATC ELPHGLATC ELPHGLREFATC -ELPHPCATC
665-0562EA 0 EUR InStock
665-0562 665-0563 664-0241 663-0376 663-0378 663-0380 663-0382 664-0240 662-2428 662-2430 662-2432 663-0375 663-0377 663-0379 663-0381 664-0239 662-2427 662-2429 662-2431
Multiparameter photometer, Water-ID® The PrimeLab 2.0
Kolorimeter Mehrparameter-Kolorimeter
The next generation of multitest Photometer, with pH - EC - TDS - ORP - Temperature measurement functionality.
  • More than 140 parameters
  • 18 wavelengths (410 to 940 nm)
  • 5.5" colour, HD touch display
  • In-build barcode/QR-code scanner
  • USB Type–C (for charging, data transfer, probe connection and 4G modem)
  • Modern connection - WiFi, USB, 4G, Bluetooth®
  • Probe connector (pH, EC, TDS, Temp and ORP)
  • Auto-calibration function with certificate (software)
High-precision readings due to 18 parallel wavelengths, WiFi–USB–Bluetooth®–GSM connection, powerful software and app, synchronised via a free cloud service, large HD touch display and the possibility to connect electrodes are just some of the features of the new PrimeLab 2.0.
While conventional photometers only perform tests at selected wavelengths, the Primelab can also receive data from 18 different wavelengths in parallel and thus covers the most important parts of the UV and IR range as well as the entire VIS range. Corresponding LEDs are positioned at both 180° and 90° to also allow NTU turbidity, PTSA and fluorescein measurements.
Very narrow peaks between 390 and 950 nm enable the most accurate readings, close to the performance of a spectrophotometer. PrimeLab 2.0 has alos the option to connect test probes, such as those used for pH, EC, TDS, Salinity, ORP and temperature.
Connnection is made by a USB-Type C cable with an A/D-exchange-box link. By connecting probes to the PrimeLab 2.0, the photometer manages the probe, obtaining readings which can be stored under the user-defined Accounts and synchronized to the LabCOM Cloud (optional).
Bestellinformation: PrimeLab 2.0 has the option to connect test probes, such as those used for pH, EC, TDS, Salinity, ORP and temperature. Connection is made by a USB-Type C cable with an A/D-exchange-box link (ProbeBOX). ProbeBOX starter kits and electrodes are sold separately.
Lieferumfang: All ProbeBOX Starter Kits are equipped generally with a grey plastic carrying case with foam insert, an A/D changer-box with BNC-in and USB-Typ-C-Out (662-2433) and a PboxCable USB-cable Typ-C / Typ-C (662-2434).
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge