pH Sensor Robust, Memosens CPL51E
Lieferant: ENDRESS HAUSER
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
CPL51E-1061/0
662-2458EA 279 EUR InStock
662-2458
pH Sensor Robust, Memosens CPL51E
Elektroden pH-Elektroden
Potentiometric pH glass sensor with plastic shaft for laboratory measurement and random sampling in the process. Connectable via inductive, digital connection with Memosens technology.
  • pH sensor for drinking water and wastewater applications
  • Polymer shaft protects the sensor from breaking
  • Open junction to minimize the risk of clogging
The Memosens 2.0 pH sensor CPL51E with a robust plastic shaft can be used both for analyzing process samples directly in the process or in the laboratory.
Thanks to Memosens technology, the sensors provide reliable, highly accurate measurements with a fast response time.
The inductive Memosens coupling between the corresponding Memosens sensor and the mobile CML18 handheld device is insensitive to moisture and corrosion.
The Memosens technology is based on a true plug-and-play principle. Just connect the sensor and you are ready to start your measurements. Measured values, calibration data and history are stored directly in the sensor. This means status values can be read out and a sensor check is also possible. An excellent feature to boost your GLP.
All sensors are supplied pre-calibrated. Thus, they are directly ready for use for your random sample measurement when the corresponding sensor is connected to the associated mobile handheld device Liquiline Mobile CML18.
Zertifizierungen: EC_00828_03.22; EU_01000_01.22; UK_00482_01.22. Nonhazardous area approval, CE + UKCA marking.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge