Cleaning and Storage Solutions
Lieferant: HAMILTON BONADUZ
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
238931
662-9223EA 32.8 EUR InStock
662-9223
Cleaning and Storage Solutions
Elektrolyte Elektrodenaufbewahrungslösungen
Hamilton storage solution is recommended to increase sensor longevity when not in use. The liquid solution is acid buffered to help with sensor response time after storage as well as prevent any biological growth.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge