Pocket conductivity/TDS testers, Thermo Scientific™ Eutech™ Elite CTS
Lieferant: EUTECH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
ELITECTSCUP ELITECTSCUPKIT ELITECTSPIN ELITECTSPINKIT
663-0357EA 160 EUR InStock
663-0357 663-0352 663-0358 663-0351
Pocket conductivity/TDS testers, Thermo Scientific™ Eutech™ Elite CTS
Elektrochemische Messgeräte Leitfähigkeitsmessgeräte
These rugged, user-friendly testers simultaneously display conductivity/TDS/salinity with temperature. They are available with a pin or cup sensor. The robust cup sensor features an industrial grade SS316 stainless steel sensor element which protects against harsh samples and ensures superior chemical durability. It also allows the user to measure smaller volumes by placing the liquid directly into the cup.
  • Intuitive keypad and large graphic LCD with backlight
  • Auto-hold readings; One-push calibration
  • Easy to replace sensor
  • Leakproof cap with fill-line indicator
  • Wide, stable base allows the tester to stand independently
  • Lanyard provision for ‘on the go’ measurements
  • IP 67 waterproof rating
Typical applications include: Environmental, agriculture, aquaculture and aquariums, hydroponics, pools and spas, education, and industrial (cooling towers).
Warranty: 1 year
Lieferumfang: Supplied with cup or pin sensor, lanyard, batteries and manual.
The kits additionally include a soft carrying case and conductivity standards (1413 µS/cm and 2764 µS/cm (5 sachets each)).
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge