Four-Electrode Conductivity Sensor Wide, Memosens CLL47E
Lieferant: ENDRESS HAUSER
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
CLL47E-1014/0
663-0383EA 914 EUR InStock
663-0383
Four-Electrode Conductivity Sensor Wide, Memosens CLL47E
Elektroden Leitfähigkeitselektroden
Four-electrode-sensor with stainless steel, ceramic and platinum for laboratory and sample measurement in the field with wide measuring range. Connectable via inductive, digital connection with Memosens technology.
  • Wide measuring range thanks to 4-electrode-sensor technology: enables analysis of many different samples
  • Solid platinum electrodes provide easy cleaning and very long lifetime
The Memosens 2.0 conductivity sensor CLL47E is a four-electrode conductivity sensor for analyzing process samples directly in the process or in the laboratory.
Thanks to Memosens technology, the sensors provide reliable, highly accurate measurements with a fast response time.
The inductive Memosens coupling between the corresponding Memosens sensor and the mobile CML18 handheld device is insensitive to moisture and corrosion.
The Memosens technology is based on a true plug-and-play principle. Just connect the sensor and you are ready to start your measurements. Measured values, calibration data and history are stored directly in the sensor. This means status values can be read out and a sensor check is also possible. An excellent feature to boost your GLP.
All sensors are supplied pre-calibrated. Thus, they are directly ready for use for your random sample measurement when the corresponding sensor is connected to the associated mobile handheld device Liquiline Mobile CML18.
Zertifizierungen: Nonhazardous area, CE + UKCA marking.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge