Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Flow-through vessel

Lieferant: WTW

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

108060 203730 302750 302760 903842
663-1801EA 455 EUR InStock
663-1801 715-2994 663-1803 WTWA302760 WTWA903842
Flow-through vessel
Reaktionsgefäße Reaktionsgefäße, Becherform
Flow vessels for WTW conductivity measuring cells are perfectly designed to perform measurements without the interference of carbon dioxide through the air or to examine the conductivity of solutions in closed circuits.

D 3Sen: Designed for everybody who must perform flow-through measurements in the field, i.e. ground water pumping tests. It consists of a rugged flow-through vessel in holder and a tripod, also suitable for pole mounting. With hose fittings for standard hoses with a diameter of 19 mm. Three places are available for pH/ORP, conductivity and dissolved oxygen sensors (CellOx/FDO). Including mounting accessories.

Achtung! Bitte beachten: Please note: The overpressure in the system must not exceed 0,5 bar.

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge