Oxygen Sensor, Memosens COS81E
Lieferant: ENDRESS HAUSER
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
COS81E-10L1/0 COS81E-14W2/0 COS81E-14Q0/0 COS81E-14V7/0
664-0243EA 2740 EUR InStock
664-0243 664-0244 664-0245 664-0246
Oxygen Sensor, Memosens COS81E
Elektroden Sauerstoff-Elektroden
Memosens 2.0 hygienic sensor for the life sciences and food industries.
  • Memosens COS81E is designed according to EHEDG and ASME BPE. It avoids cross-contamination and is fully compliant to USP class VI and FDA as well as all GMP and GLP requirements
  • High process up-time: Precalibrate the sensor in your lab and then swap it into your process with plug and play. It does not need polarization time and is immediately ready to measure
  • Reliable measurement: A built-in reference LED compensates the ageing of the measuring LED. This ensures precise measured values even after frequent CIP/SIP and autoclaving
  • Reduce your maintenance effort: Memosens COS81E does not have a difficult-to-handle electrolyte or sensitive membrane. Just exchange the sensor cap, perform a calibration and you are done
  • The sensor can be used in process applications as well as benchtop fermenters. Providing you with 100% measuring consistency from the first lab trials to the final scaled-up process and your process lab
  • IIoT ready: Memosens 2.0 offers extended storage of calibration and process data, enabling better trend identification and providing a future-proof basis for predictive maintenance and enhanced IIoT services
Memosens COS81E is the ideal optical oxygen sensor for hygienic applications. Its well-tuned measuring system prevents ageing of the optical spot and offers precise, long-term stable measurements for high product quality. With Memosens 2.0 digital technology, COS81E offers extended data storage providing the perfect basis for predictive maintenance and efficient operation.
Memosens COS81E measures oxygen in applications with high demands to sterility and hygiene such as fermentation processes, from lab to process.
Measurement range: 0 to 30 mg/L
Zertifizierungen: EN 61326-1 EN 61326-2-3; EN 50581; EN IEC 60079-0; UKCA; Nonhazardous area approval.
Zubehörinformationen: Maintenance service kit COV81 contains optical cap, O-rings and further material for maintenance of Memosens COS81E oxygen sensor.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge