68119.03
68119.06
68119.03EA
80.1
EUR
InStock
68119.03
68119.06
Dimethyldiselenid ≥99%
Dimethyl diselenide
Purity: 99%
CAS No.: 7101-31-7
Molecular Formula: (CH3)2Se2
Formula Weight: 187.99
Flash Point: 60°(140°F)
Physical Form: Liquid
Density: 1.987
Refractive Index: 1.6388
Sensitivity: Air and Moisture Sensitive
Odour: Unpleasant
Boiling Point: 155-157°
UN No.: 2929
MDL No.: MFCD00008254