Organic reference standard, Benzyl benzoate, analytical standard, Supelco®
Lieferant: SUPELCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
68183-1ML
68183-1MLEA 65 EUR InStock
68183-1ML
Organic reference standard, Benzyl benzoate, analytical standard, Supelco®
Standards Organische Standards
Benzyl benzoate undergoes hydrogenation reaction catalyzed by a system of (Ru(acac)₃) (Where Ru-ruthenium/acac-acetylacetonate) with MeC(CH₂PPh₂)₃ (tripodal triphosphine ligand).
In the presence of excess of triethylamine in trifluoroethanol obtain benzyl alcohol.
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=afb731866a56b4d1b567a01b3390d9e716fd7ab3&vid=afb731866a56b4d1b567a01b3390d9e716fd7ab3&prid=web&clid=SAPDAM
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge