Alkane standard mixture for performance tests of GC-systems, C10 - C40 (all even), analytical standard, Supelco®
Lieferant: SUPELCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
68281-2ML-F
68281-2ML-FEA 107 EUR InStock
68281-2ML-F
Alkane standard mixture for performance tests of GC-systems, C10 - C40 (all even), analytical standard, Supelco®
Standards Mineralölindustrie-Standards
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge