Organic reference standard, Drometrizole trisiloxane, analytical standard, Supelco®
Lieferant: SUPELCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
68790-50MG
68790-50MGEA 275 EUR InStock
68790-50MG
Organic reference standard, Drometrizole trisiloxane, analytical standard, Supelco®
Standards Organische Standards
Drometrizole trisiloxane (DRT) belongs to the lipophilic benzotriazole derivative class of compounds which is commonly utilised in sunscreen cosmetics.
Determined in active pharmaceutical ingredients (APIs), formulations and stability samples by reversed phase ultra-high performance liquid chromatography (RP-UPLC) coupled to electrospray ionization-quadrupole time-of-flight tandem mass spectrometry (ESI-QTOF-MS/MS), Suncare formulations by high performance liquid chromatography (HPLC) with ultraviolet (UV) detection, Cosmetic products by LC-UV and sewage sludge samples by QuEChERS extraction and gas chromatography combined with MS/MS.
Verpackung: Bottomless glass bottles are inserted with fused cone.
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a73d7c95a16176a78f2e994ca94d3677e359ce06&vid=a73d7c95a16176a78f2e994ca94d3677e359ce06&prid=web&clid=SAPDAM
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge