Vector, pET-24d(+) DNA
Lieferant: Novagen (Part of Merck)
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
69752-3
69752-3EA 291 EUR InStock
69752-3
Vector, pET-24d(+) DNA
Vektoren, Plasmide und Bibliotheken
The pET-24a-d(+) vectors carry an N-terminal T7·Tag® sequence plus an optional C-terminal His·Tag® sequence.
Lieferumfang: The pET vectors are supplied as purified plasmid DNA (10 µg). And also includes an induction control strain (supplied as a glycerol stock).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge