pET-30a(+) DNA, Novagen®
Lieferant: Novagen (Part of Merck)
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
69909-3
69909-3EA 306 EUR InStock
69909-3
pET-30a(+) DNA, Novagen®
Vektoren, Plasmide und Bibliotheken
The pET-30a-c(+) vectors carry an N-terminal His Tag®/thrombin/S Tag™/enterokinase configuration plus an optional C-terminal His tag sequence.
Lieferumfang: The pET Vectors are supplied as purified plasmid DNA (10 µg). And also includes an induction control strain (supplied as a glycerol stock).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge