Western blotting markers, Perfect Protein™, Novagen®
Lieferant: Novagen (Part of Merck)
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
69959-3
69959-3EA 115 EUR InStock
69959-3
Western blotting markers, Perfect Protein™, Novagen®
Elektrophorese-Reagenzien Molekulargewichtsmarker
The Perfect Protein™ Western Markers consist of seven recombinant S•Tag™/His•Tag® fusion proteins that can be detected on any Western blot using an S-protein AP or HRP conjugate. As an additional option, the markers can also be detected with an His•Tag Monoclonal Antibody and an AP or HRP conjugated secondary antibody. The Perfect Protein Western Markers serve as precise size standards that appear simultaneously on the blot with target proteins, eliminating the uncertainty and imprecision associated with other methods.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge