Anti-IgA Goat Polyclonal Antibody
Lieferant: Tonbo Biosciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
70-8015-M001
70-8015-M001EA 200 EUR InStock
70-8015-M001
Anti-IgA Goat Polyclonal Antibody
Antikörper
Goat anti-Human IgA (alpha chain), F(ab')2 Fragment - Affinity Pure
Type: Secondary
Antigen: IgA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: α chain
Host: Goat
Isotype:
Reactivity: Human
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge