Top Works™-Systeme für Medienflaschen-Verschlüsse, Nalgene®
Lieferant: Thermo Fisher Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
2132-1003
700-0001EA 380 EUR InStock
700-0001
Top Works™-Systeme für Medienflaschen-Verschlüsse, Nalgene®
Verschlüsse Flaschenverschlüsse
PSF, closures with platinum cured silicone inserts.
  • They fit the corresponding threads on bottles by Schott, Corning and Wheaton
  • For thread size GL 45
  • Autoclavable, leakproof
These closures make it easier to transfer liquids aseptically from most conventional glass media bottles.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge