Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Dual-gel electrophoresis system, Owl™ P8DS

Lieferant: THERMO OWL SCIENTIFIC

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

P8DS-1 P8DS-2 P8DS-3
730-0315EA 2250 EUR InStock
730-0315 700-2340 700-2341
Dual-gel electrophoresis system, Owl™ P8DS
Elektrophorese-Systeme Vertikale Gelelektrophorese-Systeme
The Owl™ P9DS dual gel system is designed for quick and easy casting and running of one or two gels on the same device. When used with the available combs, spacers, alumina plates and glass, this system is an ideal tool for second dimension protocols of 2-D electrophoresis, SDS and agarose gels, and native PAGE electrophoresis.

  • Agarose plug method makes gel casting simple and quick
  • Gel assemblies do not have to be moved once set up
  • Two individual steps of casting and running are combined
  • Simple clamping mechanism makes loading glass cassettes, and most precast gels, trouble free

Gel W×H: 100×80 to 100 mm
Footprint W×H×D: 170×140×130 mm
Running buffer volume: 300 ml (upper chamber 150 ml, lower chamber 150 ml)

Lieferumfang: Complete system includes upper buffer chamber with internal cooling core, lower buffer chamber, SuperSafe lid with attached power leads, 4 blank glass plates, 2 notched glass plates, 2 notched alumina plates, 2 combs (10 well, 1,5 mm thick), spacer set (0,8 mm thick), spacer placer, and blocking plate for single gel operation.

Order Now
 

PRODUKT

TECHNISCHE DATEN

ZUBEHÖR

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge