COD photometric test kits, Test’n Tube
Lieferant: HACH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
2125851 2125951 2415951 2415851 2345952 2345852 2623451
705-0096EA 119 EUR InStock
705-0096 705-0097 705-0129 705-0128 HACH2345952 HACH2345852 705-2048
COD photometric test kits, Test’n Tube
Umwelt-Prüfkits
Cuvette Test complete with all reagents, optimised for simple handling on the colorimeters DR900 and DR890 and DR850*.
  • Chromsulphuric acid method
Achtung! Bitte beachten: *705-0096 is not suitable for DR 850
**705-0128 is only suitable for the DR 4000 and the DR 2500
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge