Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Generic pocket colorimeters, DR300

Lieferant: Hach

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

LPV4459960110 LPV4459950110 LPV4459952110 LPV4459965110
705-1179EA 849 EUR InStock
705-1179 705-1177 705-1178 705-1180
Generic pocket colorimeters, DR300
Kolorimeter Einzelparameter-Kolorimeter
Generic pocket colorimeter DR300 instruments are intended for customised applications not included in standard analysis systems. They are available in a range of wavelengths appropriate for many custom calibrations. In addition to the factory absorbance calibration (0 to 2,5 A), the instruments offer two user-entered, two-point linear calibrations, and provide a choice of either 0,00 to 2,55 or 0,0 to 99,0 for the display value.

  • Single parameter
  • Waterproof, lightweight, rugged
  • Backlit display
  • Data logging recall function

These instruments accept two user-prepared calibration curves. The calibration curve can be from 0 to 2,5 absorbance. Make sure that the calibration curve includes standard values that are less and more than the range of interest. The instrument range will be the same as the calibration range. For example, when the standards that are used are 1,00; 2,00 and 4,00, the instrument range is 1,00 to 4,00. There are two options to enter a user calibration curve:
- By standards: Standard solution values are entered with the keypad and the absorbance values are measured
- By data entered via keypad: Standard solution values and absorbance values are entered using the keypad.

Bestellinformation: Reagents must be purchased separately.

Lieferumfang: Each DR300 pocket colorimeter is delivered as an instrument kit in a sturdy custom carrying case, including batteries, sample cells (1" Ø and 10 mm Ø, two of each), and a printed user manual.

Order Now
 

PRODUKT

TECHNISCHE DATEN

ZUBEHÖR

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge