BugBuster® Ni-NTA His•Bind® purification kit
Lieferant: Novagen (Part of Merck)
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
70793-3
70793-3EA 716 EUR InStock
70793-3
BugBuster® Ni-NTA His•Bind® purification kit
Protein-Aufbereitungs Protein Purification Kits
BugBuster®Ni-NTA HIS-BIND® Purification Kit is used for protein purification. Ni-NTA HIS-BIND® Resin is high-performance Ni2+-charged agarose used for rapid, one-step purification of proteins containing a polyhistidine tag sequence.
BugBuster®Ni-NTA HIS-BIND® Purification Kit has been used for the purification of His tagged proteins such as trehalose-6-phosphate phosphatase (TPP) , Ras(WT) proteins and CsgA (major curlin subunit) proteins.
Composants
2 × 100 mlBugBuster Protein Extraction Reagent
10,000 UBenzonase Nuclease, purity >90%
10 mlNi-NTA His•Bind Resin
pkg/4Chromatography Columns
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge